Phosphoric acid, for analysis, 85 wt% solution in water, Thermo Scientific Chemicals
Phosphoric acid, for analysis, 85 wt% solution in water, Thermo Scientific Chemicals
Phosphoric acid, for analysis, 85 wt% solution in water, Thermo Scientific Chemicals
Phosphoric acid, for analysis, 85 wt% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phosphoric acid, for analysis, 85 wt% solution in water, Thermo Scientific Chemicals
CAS: 7664-38-2 | H3O4P | 97.994 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
1 L
Packaging:
Glass bottle
Catalog number FSA201140025
Price (MYR)
911.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
911.00
Add to cart
Chemical Identifiers
CAS145224-92-6
IUPAC Namesodium (4R)-4-[(1R,3aS,3bR,5aR,7R,9aS,9bS,11S,11aR)-7,11-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate hydrate
Molecular FormulaC24H41NaO5
InChI KeyNDVHNZISGVSFMP-WTCAICSISA-M
SMILESO.[Na+].[H][C@@]1(CC[C@@]2([H])[C@]3([H])CC[C@]4([H])C[C@H](O)CC[C@]4(C)[C@@]3([H])C[C@H](O)[C@]12C)[C@H](C)CCC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Optical Rotation44.0 ± 2.0° (C=2 in water, UK-sourced material)
FormPowder
Assay from Supplier's CofA≥97.5% (HPLC, US-sourced material)
Identification (FTIR)Conforms (UK-sourced material)