Phosphoric acid, for analysis, 85 wt% solution in water, Thermo Scientific Chemicals
Phosphoric acid, for analysis, 85 wt% solution in water, Thermo Scientific Chemicals
Phosphoric acid, for analysis, 85 wt% solution in water, Thermo Scientific Chemicals
Phosphoric acid, for analysis, 85 wt% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phosphoric acid, for analysis, 85 wt% solution in water, Thermo Scientific Chemicals
CAS: 7664-38-2 | H3O4P | 97.994 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number FSA201140010
Price (MYR)
539.00
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
539.00
Add to cart
Chemical Identifiers
CAS4584-49-0
IUPAC Namehydrogen (2-chloropropyl)dimethylamine chloride
Molecular FormulaC5H13Cl2N
InChI KeyOCWGRWAYARCRTQ-UHFFFAOYNA-N
SMILES[H+].[Cl-].CC(Cl)CN(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=98.0 % (On dry substance)
Appearance (Form)Crystalline powder
Appearance (Color)White to cream
Infrared spectrumConforms
Loss on drying=<1 %