Gallium(III) oxide, 99.99+%, (trace metal basis), Thermo Scientific Chemicals
Gallium(III) oxide, 99.99+%, (trace metal basis), Thermo Scientific Chemicals
Gallium(III) oxide, 99.99+%, (trace metal basis), Thermo Scientific Chemicals
Gallium(III) oxide, 99.99+%, (trace metal basis), Thermo Scientific Chemicals
Gallium(III) oxide, 99.99+%, (trace metal basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Gallium(III) oxide, 99.99+%, (trace metal basis), Thermo Scientific Chemicals
CAS: 12024-21-4 | Ga2O3 | 187.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Catalog number FSA201100050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS100-83-4
IUPAC Name3-hydroxybenzaldehyde
Molecular FormulaC7H6O2
InChI KeyIAVREABSGIHHMO-UHFFFAOYSA-N
SMILESOC1=CC=CC(C=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Assay (GC)≥96.0%
Melting Point (clear melt)101-106?C
Appearance (Color)White to cream to yellow to orange to pale brown
FormCrystalline powder or powder
Suitable for battery materials development.
for ultra trace analysis
RUO – Research Use Only