Phosphorus pentoxide, 98%, extra pure, Thermo Scientific Chemicals
Phosphorus pentoxide, 98%, extra pure, Thermo Scientific Chemicals
Phosphorus pentoxide, 98%, extra pure, Thermo Scientific Chemicals
Phosphorus pentoxide, 98%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phosphorus pentoxide, 98%, extra pure, Thermo Scientific Chemicals
CAS: 1314-56-3 | O5P2 | 141.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Catalog number FSA200892500
Price (MYR)
360.00
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
360.00
Add to cart
Chemical Identifiers
CAS206551-43-1
IUPAC Name(5-acetylthiophen-2-yl)boronic acid
Molecular FormulaC6H7BO3S
InChI KeyDCNMATSPQKWETQ-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=C(S1)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale brown to brown
FormPowder
Assay (Aqueous acid-base Titration)≥97.5%
Proton NMRConforms to structure
Assay (HPLC)≥97.5%
View more
Adsorbent
RUO – Research Use Only