Chrysoidin, pure
Chrysoidin, pure
Chrysoidin, pure
Thermo Scientific Chemicals
Chrysoidin, pure
CAS: 532-82-1 | C12H12N4·HCl | 248.72 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA20074050050 gGlass bottle
Catalog number FSA200740500
Price (MYR)
403.00
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
403.00
Add to cart
Specifications
Chemical Name or MaterialChrysoidin
CAS532-82-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes skin irritation.
Suspected of causing genetic defects.
Causes serious eye damage.
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective