Rhodium on alumina, 5% Rh, powder, Thermo Scientific Chemicals
Rhodium on alumina, 5% Rh, powder, Thermo Scientific Chemicals
Rhodium on alumina, 5% Rh, powder, Thermo Scientific Chemicals
Rhodium on alumina, 5% Rh, powder, Thermo Scientific Chemicals
Rhodium on alumina, 5% Rh, powder, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Catalog number FSA199570050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS58052-80-5
IUPAC Name2,4,6-trimethyl-1,3,5-triazinane-1,3,5-triium
Molecular FormulaC6H18N3
InChI KeyMZSSRMMSFLVKPK-UHFFFAOYSA-Q
SMILESCC1[NH2+]C(C)[NH2+]C(C)[NH2+]1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to light beige
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Titration with HClO497.5 to 102.5 %