Fusaric acid, 99%, Thermo Scientific Chemicals
Fusaric acid, 99%, Thermo Scientific Chemicals
Fusaric acid, 99%, Thermo Scientific Chemicals
Fusaric acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Fusaric acid, 99%, Thermo Scientific Chemicals
CAS: 536-69-6 | C10H13NO2 | 179.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Catalog number FSA198962500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS33574-02-6
IUPAC Name(iodomethyl)cyclopropane
Molecular FormulaC4H7I
InChI KeyJHKJQWFHNIOUKY-UHFFFAOYSA-N
SMILESICC1CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to orange
Assay (GC)≥94.0%
Refractive Index1.5385-1.5445 @ 20?C