Boron trichloride, 1M solution in hexane, AcroSeal™, Thermo Scientific Chemicals
Boron trichloride, 1M solution in hexane, AcroSeal™, Thermo Scientific Chemicals
Boron trichloride, 1M solution in hexane, AcroSeal™, Thermo Scientific Chemicals
Boron trichloride, 1M solution in hexane, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Boron trichloride, 1M solution in hexane, AcroSeal™, Thermo Scientific Chemicals
CAS: 10294-34-5 | BCl3 | 117.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
800 mL
100 mL
Packaging:
View all
AcroSeal™ Glass bottle
Catalog number FSA198928000
Price (MYR)
3,021.00
Add to cart
Quantity:
800 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (MYR)
3,021.00
Add to cart
Chemical Identifiers
CAS25322-68-3
IUPAC Nameethane-1,2-diol
Molecular FormulaC2H6O2
InChI KeyLYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILESOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Assay (unspecified)Viscosity: 30-50 mPa sec
FormPowder
CommentMolecular Weight: 100,000 (approx.)
Lewis acid
RUO – Research Use Only