Boron trichloride, 1M solution in hexane, AcroSeal™, Thermo Scientific Chemicals
Boron trichloride, 1M solution in hexane, AcroSeal™, Thermo Scientific Chemicals
Boron trichloride, 1M solution in hexane, AcroSeal™, Thermo Scientific Chemicals
Boron trichloride, 1M solution in hexane, AcroSeal™, Thermo Scientific Chemicals
Boron trichloride, 1M solution in hexane, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Boron trichloride, 1M solution in hexane, AcroSeal™, Thermo Scientific Chemicals
CAS: 10294-34-5 | BCl3 | 117.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
View all
AcroSeal™ Glass Bottle
Catalog number FSA198921000
Price (MYR)
513.00
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
513.00
Add to cart
Chemical Identifiers
CAS193978-23-3
IUPAC Name4,4,5,5-tetramethyl-2-(thiophen-2-yl)-1,3,2-dioxaborolane
Molecular FormulaC10H15BO2S
InChI KeyFFZHICFAHSDFKZ-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=CC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder or crystals or crystalline powder
Melting Point64.0-71.0°C
Assay from Supplier's CofA≥97.5%
Lewis acid
RUO – Research Use Only