Selenious acid, 98%, Thermo Scientific Chemicals
Selenious acid, 98%, Thermo Scientific Chemicals
Selenious acid, 98%, Thermo Scientific Chemicals
Selenious acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Selenious acid, 98%, Thermo Scientific Chemicals
CAS: 7783-00-8 | H2O3Se | 128.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Glass bottle
Catalog number FSA198870500
Price (MYR)
412.00
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
412.00
Add to cart
Specifications
Chemical Name or MaterialSelenious acid
ColorPink or White
CAS7783-00-8
Melting Point70.0°C
Health Hazard 1GHS Signal Word: Danger