2,2-Dimethyl-1,3-dioxane-4,6-dione, 98%, Thermo Scientific Chemicals
2,2-Dimethyl-1,3-dioxane-4,6-dione, 98%, Thermo Scientific Chemicals
2,2-Dimethyl-1,3-dioxane-4,6-dione, 98%, Thermo Scientific Chemicals
2,2-Dimethyl-1,3-dioxane-4,6-dione, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,2-Dimethyl-1,3-dioxane-4,6-dione, 98%, Thermo Scientific Chemicals
CAS: 2033-24-1 | C6H8O4 | 144.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA198450250
Price (MYR)
366.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
366.00
Add to cart
Chemical Identifiers
CAS555-44-2
IUPAC Name1,3-bis(hexadecanoyloxy)propan-2-yl hexadecanoate
Molecular FormulaC51H98O6
InChI KeyPVNIQBQSYATKKL-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)White to almost white
Infrared spectrumConforms
HPLC>=98.5 %