Periodic acid, 99+%, extra pure, Thermo Scientific Chemicals
Periodic acid, 99+%, extra pure, Thermo Scientific Chemicals
Periodic acid, 99+%, extra pure, Thermo Scientific Chemicals
Periodic acid, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Periodic acid, 99+%, extra pure, Thermo Scientific Chemicals
Periodic acid, 99+%, H5IO6, CAS Number-10450-60-9, pentahydroxidooxidoiodine, paraperiodic acid, orthoperiodic acid, acid, periodic, hexaoxoiodic 5-acid, tetrahydroxyiodous acid, pentahydroxy-l5-iodane oxide, periodic acid, periodic acid, cryst, unii-ak1d44l87g, 100g, 1.0% max. Iodic Acid
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Catalog number FSA198401000
Price (MYR)
808.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
808.00
Add to cart
Chemical Identifiers
CAS3734-33-6
IUPAC Namebenzyl({[(2,6-dimethylphenyl)carbamoyl]methyl})diethylazanium benzoate
Molecular FormulaC28H34N2O3
InChI KeyVWTINHYPRWEBQY-UHFFFAOYSA-N
SMILES[O-]C(=O)C1=CC=CC=C1.CC[N+](CC)(CC(=O)NC1=C(C)C=CC=C1C)CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystalline powder
Assay (unspecified)99.5-101.0%
Residue on ignition0.1% max.
Loss on Drying3.0% max.