Periodic acid, 99+%, extra pure, Thermo Scientific Chemicals
Periodic acid, 99+%, extra pure, Thermo Scientific Chemicals
Periodic acid, 99+%, extra pure, Thermo Scientific Chemicals
Periodic acid, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Periodic acid, 99+%, extra pure, Thermo Scientific Chemicals
Periodic acid, 99+%, H5IO6, CAS Number-10450-60-9, pentahydroxidooxidoiodine, paraperiodic acid, orthoperiodic acid, acid, periodic, hexaoxoiodic 5-acid, tetrahydroxyiodous acid, pentahydroxy-l5-iodane oxide, periodic acid, periodic acid, cryst, unii-ak1d44l87g, 100g, 1.0% max. Iodic Acid
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Catalog number FSA198400025
Price (MYR)
8,283.00
Add to cart
Quantity:
2.5 kg
Request bulk or custom format
Price (MYR)
8,283.00
Add to cart
Chemical Identifiers
CAS109-01-3
IUPAC Name1-methylpiperazine
Molecular FormulaC5H12N2
InChI KeyPVOAHINGSUIXLS-UHFFFAOYSA-N
SMILESCN1CCNCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<20 APHA
Infrared spectrumConforms
Refractive index1.4645 to 1.4665 (20°C, 589 nm)
GC>=98.5 %