n-Hexane, 95+%, extra pure, Thermo Scientific Chemicals
n-Hexane, 95+%, extra pure, Thermo Scientific Chemicals
n-Hexane, 95+%, extra pure, Thermo Scientific Chemicals
n-Hexane, 95+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Hexane, 95+%, extra pure, Thermo Scientific Chemicals
CAS: 110-54-3 | C6H14 | 86.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 L
25 L
Packaging:
View all
Plastic Drum
Catalog number FSA197360050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 L
Packaging:
Plastic Drum
Request bulk or custom format
Chemical Identifiers
CAS2312-23-4
IUPAC Namehydrogen (3-chlorophenyl)hydrazine chloride
Molecular FormulaC6H8Cl2N2
InChI KeyCRRIAWUJYMLJOE-UHFFFAOYSA-N
SMILES[H+].[Cl-].NNC1=CC=CC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to pale brown
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥96.0 to ≤104.0%