Molecular sieves 4A, 8 to 12 mesh, Thermo Scientific Chemicals
Molecular sieves 4A, 8 to 12 mesh, Thermo Scientific Chemicals
Molecular sieves 4A, 8 to 12 mesh, Thermo Scientific Chemicals
Molecular sieves 4A, 8 to 12 mesh, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Molecular sieves 4A, 8 to 12 mesh, Thermo Scientific Chemicals
CAS: 70955-01-0 | C20H25FN2O8 | 440.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 kg
Packaging:
View all
Glass bottle
Catalog number FSA197275000
Price (MYR)
695.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
695.00
Add to cart
Chemical Identifiers
CAS121-39-1
IUPAC Nameethyl 3-phenyloxirane-2-carboxylate
Molecular FormulaC11H12O3
InChI KeyGOMAKLPNAAZVCJ-UHFFFAOYNA-N
SMILESCCOC(=O)C1OC1C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to yellow
Assay (GC)≥90.0%
Refractive Index1.4785-1.5585 @ 20?C
Drying agent
RUO – Research Use Only