Molecular sieves 4A, 8 to 12 mesh, Thermo Scientific Chemicals
Molecular sieves 4A, 8 to 12 mesh, Thermo Scientific Chemicals
Molecular sieves 4A, 8 to 12 mesh, Thermo Scientific Chemicals
Molecular sieves 4A, 8 to 12 mesh, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Molecular sieves 4A, 8 to 12 mesh, Thermo Scientific Chemicals
CAS: 70955-01-0 | C20H25FN2O8 | 440.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
500 g
Packaging:
View all
Plastic drum
Catalog number FSA197270050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Specifications
Bulk Density680 to 760g/L
Chemical Name or MaterialMolecular sieves 4A
ColorBeige
PackagingPlastic drum
Residual Water1.5% max.
View more
Drying agent
RUO – Research Use Only