Cobalt(II) acetate tetrahydrate, 98-102%, ACS reagent
Cobalt(II) acetate tetrahydrate, 98-102%, ACS reagent
Cobalt(II) acetate tetrahydrate, 98-102%, ACS reagent
Cobalt(II) acetate tetrahydrate, 98-102%, ACS reagent
Thermo Scientific Chemicals
Cobalt(II) acetate tetrahydrate, 98-102%, ACS reagent
CAS: 6147-53-1 | C4H6CoO4·4H2O | 249.08 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA197101000100 gPlastic bottle
Catalog number FSA197101000
Price (MYR)
712.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
712.00
Add to cart
Chemical Identifiers
CAS4111-54-0
IUPAC Namelithium(1+) bis(propan-2-yl)azanide
Molecular FormulaC6H14LiN
InChI KeyZCSHNCUQKCANBX-UHFFFAOYSA-N
SMILES[Li+].CC(C)[N-]C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark yellow to orange to dark red-brown
AppearanceIf precipitation is present, gently mix
Titration with HClO424.6 to 30.0 %
Appearance (Form)Clear to slightly hazy solution
Appearanceatmosphere