Polyphosphoric acid, pure, >84% phosphate (as P2O5), Thermo Scientific Chemicals
Polyphosphoric acid, pure, &gt;84% phosphate (as P<sub>2</sub>O<sub>5</sub>), Thermo Scientific Chemicals
Polyphosphoric acid, pure, &gt;84% phosphate (as P<sub>2</sub>O<sub>5</sub>), Thermo Scientific Chemicals
Polyphosphoric acid, pure, &gt;84% phosphate (as P<sub>2</sub>O<sub>5</sub>), Thermo Scientific Chemicals
Polyphosphoric acid, pure, &gt;84% phosphate (as P<sub>2</sub>O<sub>5</sub>), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Polyphosphoric acid, pure, >84% phosphate (as P2O5), Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
1 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number FSA196950025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS7154-73-6
IUPAC Name2-(pyrrolidin-1-yl)ethan-1-amine
Molecular FormulaC6H14N2
InChI KeyWRXNJTBODVGDRY-UHFFFAOYSA-N
SMILESNCCN1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4677 to 1.4697 (20°C, 589 nm)
Appearance (Color)Clear colorless to light brown