Polyphosphoric acid, pure, >84% phosphate (as P2O5), Thermo Scientific Chemicals
Polyphosphoric acid, pure, &gt;84% phosphate (as P<sub>2</sub>O<sub>5</sub>), Thermo Scientific Chemicals
Polyphosphoric acid, pure, &gt;84% phosphate (as P<sub>2</sub>O<sub>5</sub>), Thermo Scientific Chemicals
Polyphosphoric acid, pure, &gt;84% phosphate (as P<sub>2</sub>O<sub>5</sub>), Thermo Scientific Chemicals
Polyphosphoric acid, pure, &gt;84% phosphate (as P<sub>2</sub>O<sub>5</sub>), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Polyphosphoric acid, pure, >84% phosphate (as P2O5), Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number FSA196950010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPolyphosphoric acid
CAS8017-16-1
Boiling Point550.0°C
Density2.0500g/mL
Recommended StorageMay darken over time