Fluoroboric acid, 50 wt.% solution in water, Thermo Scientific Chemicals
Fluoroboric acid, 50 wt.% solution in water, Thermo Scientific Chemicals
Fluoroboric acid, 50 wt.% solution in water, Thermo Scientific Chemicals
Fluoroboric acid, 50 wt.% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Fluoroboric acid, 50 wt.% solution in water, Thermo Scientific Chemicals
CAS: 16872-11-0 | BF4H | 87.812 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA196700250
Price (MYR)
386.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
386.00
Add to cart
Chemical Identifiers
CAS9003-20-7
Molecular Formula(C4H6O2)n
SMILESCC(=O)OC(-*)C-*
MDL NumberMFCD00084457
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear
Loss on drying=<5 %
Appearance (Form)Beads
Infrared spectrumConforms
C-X bond formation reagent
RUO – Research Use Only