Fluoroboric acid, 50 wt.% solution in water, Thermo Scientific Chemicals
Fluoroboric acid, 50 wt.% solution in water, Thermo Scientific Chemicals
Fluoroboric acid, 50 wt.% solution in water, Thermo Scientific Chemicals
Fluoroboric acid, 50 wt.% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Fluoroboric acid, 50 wt.% solution in water, Thermo Scientific Chemicals
CAS: 16872-11-0 | BF4H | 87.812 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
25 g
500 g
Packaging:
Plastic bottle
Catalog number FSA196700025
Price (MYR)
2,209.00
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
2,209.00
Add to cart
Chemical Identifiers
CAS9002-18-0
IUPAC Name2-({4-hydroxy-3-methyl-2,6-dioxabicyclo[3.2.1]octan-8-yl}oxy)-6-(hydroxymethyl)-4-methoxyoxane-3,5-diol
Molecular FormulaC14H24O9
InChI KeyGYYDPBCUIJTIBM-UHFFFAOYNA-N
SMILESCOC1C(O)C(CO)OC(OC2C3COC2C(O)C(C)O3)C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to light yellow to beige
Ash=<5 %
Solubility(1.5 % in hot water) Clear to slightly
Infrared spectrumConforms
pH6.0 to 7.5 (10 % suspension)
View more
C-X bond formation reagent
RUO – Research Use Only