Copper, 99%, powder, max. 106μm (-140 mesh), Thermo Scientific Chemicals
Copper, 99%, powder, max. 106μm (-140 mesh), Thermo Scientific Chemicals
Copper, 99%, powder, max. 106μm (-140 mesh), Thermo Scientific Chemicals
Copper, 99%, powder, max. 106μm (-140 mesh), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Copper, 99%, powder, max. 106μm (-140 mesh), Thermo Scientific Chemicals
CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
Packaging:
Glass bottle
Catalog number FSA196575000
Price (MYR)
464.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
464.00
Add to cart
Chemical Identifiers
CAS25322-68-3
IUPAC Nameethane-1,2-diol
Molecular FormulaC2H6O2
InChI KeyLYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILESOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)Clear colorless
FormLiquid
pH(5% aqueous soln.) : 4.5-7.5%
Refractive Index1.4580-1.4620 @ 20?C