Zinc standard solution, for AAS, 1 mg/ml Zn in 2-5% HCl
Zinc standard solution, for AAS, 1 mg/ml Zn in 2-5% HCl
Zinc standard solution, for AAS, 1 mg/ml Zn in 2-5% HCl
Zinc standard solution, for AAS, 1 mg/ml Zn in 2-5% HCl
Thermo Scientific Chemicals
Zinc standard solution, for AAS, 1 mg/ml Zn in 2-5% HCl
CAS: 7440-66-6 | Zn | 65.38 g/mol
Have Questions?
Catalog NumberQuantity
FSA196465000500 mL
Catalog number FSA196465000
Price (MYR)
377.00
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
377.00
Add to cart
Chemical Identifiers
CAS1314-36-9
IUPAC Namediyttrium(3+) trioxidandiide
Molecular FormulaO3Y2
InChI KeyRUDFQVOCFDJEEF-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Y+3].[Y+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
for atomic adsorption spectroscopy
RUO – Research Use Only