Sodium standard solution, for AAS, 1 mg/ml Na in 0.5N HNO3
Sodium standard solution, for AAS, 1 mg/ml Na in 0.5N HNO3
Sodium standard solution, for AAS, 1 mg/ml Na in 0.5N HNO3
Sodium standard solution, for AAS, 1 mg/ml Na in 0.5N HNO3
Thermo Scientific Chemicals
Sodium standard solution, for AAS, 1 mg/ml Na in 0.5N HNO3
CAS: 7631-99-4 | NNaO3 | 84.99 g/mol
Have Questions?
Catalog NumberQuantity
FSA196311000100 mL
Catalog number FSA196311000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS7631-99-4, 7697-37-2, 7732-18-5
Specifications
Chemical Name or MaterialSodium standard solution
Name Note1 mg/ml Na in 0.5N HNO3
PackagingPlastic bottle
CAS7631-99-4
Quantity100 mL
View more
for atomic adsorption spectroscopy
RUO – Research Use Only