Phosphorus standard solution, for AAS, 1 mg/ml P in water
Phosphorus standard solution, for AAS, 1 mg/ml P in water
Phosphorus standard solution, for AAS, 1 mg/ml P in water
Phosphorus standard solution, for AAS, 1 mg/ml P in water
Thermo Scientific Chemicals
Phosphorus standard solution, for AAS, 1 mg/ml P in water
CAS: 7723-14-0 | P | 30.97 g/mol
Have Questions?
Catalog NumberQuantity
FSA196185000500 mL
Catalog number FSA196185000
Price (MYR)
585.00
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
585.00
Add to cart
Chemical Identifiers
CAS24677-78-9
IUPAC Name2,3-dihydroxybenzaldehyde
Molecular FormulaC7H6O3
InChI KeyIXWOUPGDGMCKGT-UHFFFAOYSA-N
SMILESOC1=CC=CC(C=O)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to yellow-green or cream to brown or grey
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Clarity5% w/v in methanol: clear solution
Melting Point (clear melt)103-109?C
View more
For atomic adsorption spectroscopy
RUO – Research Use Only