Manganese standard solution, for AAS, 1mg/mL Mn in 2 to 5% HNO3, Thermo Scientific Chemicals
Manganese standard solution, for AAS, 1mg/mL Mn in 2 to 5% HNO<sub>3</sub>, Thermo Scientific Chemicals
Manganese standard solution, for AAS, 1mg/mL Mn in 2 to 5% HNO<sub>3</sub>, Thermo Scientific Chemicals
Manganese standard solution, for AAS, 1mg/mL Mn in 2 to 5% HNO<sub>3</sub>, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Manganese standard solution, for AAS, 1mg/mL Mn in 2 to 5% HNO3, Thermo Scientific Chemicals
CAS: 7439-96-5 | Mn | 54.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number FSA196115000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS1333-82-0
IUPAC Nametrioxochromium
Molecular FormulaCrO3
InChI KeyWGLPBDUCMAPZCE-UHFFFAOYSA-N
SMILESO=[Cr](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Iodometric98.5 to 101.5 %
Appearance (Color)Red to dark red to purple
Appearance (Form)Crystals or granules or flakes or prills
for atomic adsorption spectroscopy
RUO – Research Use Only