Magnesium standard solution, for AAS, 1 mg/ml Mg in 0.5N HNO3
Magnesium standard solution, for AAS, 1 mg/ml Mg in 0.5N HNO3
Magnesium standard solution, for AAS, 1 mg/ml Mg in 0.5N HNO3
Magnesium standard solution, for AAS, 1 mg/ml Mg in 0.5N HNO3
Thermo Scientific Chemicals
Magnesium standard solution, for AAS, 1 mg/ml Mg in 0.5N HNO3
CAS: 7439-95-4 | Mg | 24.30 g/mol
Have Questions?
Catalog NumberQuantity
FSA196105000500 mL
Catalog number FSA196105000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS6131-99-3
IUPAC Namesodium dimethylarsinate
Molecular FormulaC2H6AsNaO2
InChI KeyIHQKEDIOMGYHEB-UHFFFAOYSA-M
SMILES[Na+].C[As](C)([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Non-aqueous acid-base Titration)≥98.0 to ≤102.0%
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
For atomic adsorption spectroscopy
RUO – Research Use Only