Hafnium standard solution, for AAS, 1 mg/ml Hf in 5% HF
Hafnium standard solution, for AAS, 1 mg/ml Hf in 5% HF
Hafnium standard solution, for AAS, 1 mg/ml Hf in 5% HF
Hafnium standard solution, for AAS, 1 mg/ml Hf in 5% HF
Thermo Scientific Chemicals
Hafnium standard solution, for AAS, 1 mg/ml Hf in 5% HF
CAS: 7440-58-6 | Hf | 178.49 g/mol
Have Questions?
Catalog NumberQuantity
FSA196011000100 mL
Catalog number FSA196011000
Price (MYR)
364.00
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
364.00
Add to cart
Chemical Identifiers
CAS7790-69-4
IUPAC Namelithium(1+) nitrate
Molecular FormulaLiNO3
InChI KeyIIPYXGDZVMZOAP-UHFFFAOYSA-N
SMILES[Li+].[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder and/or granules
Assay (special)?2.5 % of Theoretical value (Li)
Total Metal Impurities≤0.001% (10 ppm)
Appearance (Color)White
For atomic adsorption spectroscopy
RUO – Research Use Only