Copper standard solution, for AAS, 1 mg/ml Cu in 2% HNO3, Thermo Scientific Chemicals
Copper standard solution, for AAS, 1 mg/ml Cu in 2% HNO3, Thermo Scientific Chemicals
Copper standard solution, for AAS, 1 mg/ml Cu in 2% HNO3, Thermo Scientific Chemicals
Copper standard solution, for AAS, 1 mg/ml Cu in 2% HNO3, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Copper standard solution, for AAS, 1 mg/ml Cu in 2% HNO3, Thermo Scientific Chemicals
CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number FSA195935000
Price (MYR)
562.00
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
562.00
Add to cart
Chemical Identifiers
CAS1125-88-8
IUPAC Name(dimethoxymethyl)benzene
Molecular FormulaC9H12O2
InChI KeyHEVMDQBCAHEHDY-UHFFFAOYSA-N
SMILESCOC(OC)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=97.5 %
Appearance (Color)Clear colorless
Refractive index1.4910 to 1.4940 (20°C, 589 nm)
Appearance (Form)Liquid
For atomic adsorption spectroscopy
RUO – Research Use Only