Copper standard solution, for AAS, 1 mg/ml Cu in 2% HNO3, Thermo Scientific Chemicals
Copper standard solution, for AAS, 1 mg/ml Cu in 2% HNO3, Thermo Scientific Chemicals
Copper standard solution, for AAS, 1 mg/ml Cu in 2% HNO3, Thermo Scientific Chemicals
Copper standard solution, for AAS, 1 mg/ml Cu in 2% HNO3, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Copper standard solution, for AAS, 1 mg/ml Cu in 2% HNO3, Thermo Scientific Chemicals
CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number FSA195931000
Price (MYR)
284.00
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
284.00
Add to cart
Chemical Identifiers
CAS7440-50-8, 7697-37-2, 7732-18-5
Specifications
Chemical Name or MaterialCopper standard solution
Name Note1 mg/ml Cu in 2% HNO3
PackagingPlastic bottle
CAS7440-50-8
Quantity100 mL
View more
For atomic adsorption spectroscopy
RUO – Research Use Only