Cobalt standard solution, for AAS, 1mg/mL Co in 2% HNO3, Thermo Scientific Chemicals
Cobalt standard solution, for AAS, 1mg/mL Co in 2% HNO<sub>3</sub>, Thermo Scientific Chemicals
Cobalt standard solution, for AAS, 1mg/mL Co in 2% HNO<sub>3</sub>, Thermo Scientific Chemicals
Cobalt standard solution, for AAS, 1mg/mL Co in 2% HNO<sub>3</sub>, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cobalt standard solution, for AAS, 1mg/mL Co in 2% HNO3, Thermo Scientific Chemicals
CAS: 10141-05-6 | CoN2O6 | 182.941 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number FSA195921000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS7440-67-7
IUPAC Namezirconium
Molecular FormulaZr
InChI KeyQCWXUUIWCKQGHC-UHFFFAOYSA-N
SMILES[Zr]
View more
SpecificationsSpecification SheetSpecification Sheet
Trace MetalTotal metal impurities less than 0.2%
For atomic adsorption spectroscopy
RUO – Research Use Only