Calcium standard solution, for AAS, 1mg/mL Ca in 0.5N HNO3, Thermo Scientific Chemicals
Calcium standard solution, for AAS, 1mg/mL Ca in 0.5N HNO<sub>3</sub>, Thermo Scientific Chemicals
Calcium standard solution, for AAS, 1mg/mL Ca in 0.5N HNO<sub>3</sub>, Thermo Scientific Chemicals
Calcium standard solution, for AAS, 1mg/mL Ca in 0.5N HNO<sub>3</sub>, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Calcium standard solution, for AAS, 1mg/mL Ca in 0.5N HNO3, Thermo Scientific Chemicals
CAS: 7440-70-2 | Ca | 40.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number FSA195881000
Price (MYR)
205.00
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
205.00
Add to cart
Chemical Identifiers
CAS7440-70-2, 7697-37-2, 7732-18-5
Specifications
Chemical Name or MaterialCalcium standard solution
Name Note1 mg/ml Ca in 0.5N HNO3
PackagingPlastic bottle
CAS7440-70-2
Quantity100 mL
View more
for atomic adsorption spectroscopy
RUO – Research Use Only