Cadmium standard solution, for AAS, 1mg/mL Cd in 0.5N HNO3, Thermo Scientific Chemicals
Cadmium standard solution, for AAS, 1mg/mL Cd in 0.5N HNO<sub>3</sub>, Thermo Scientific Chemicals
Cadmium standard solution, for AAS, 1mg/mL Cd in 0.5N HNO<sub>3</sub>, Thermo Scientific Chemicals
Cadmium standard solution, for AAS, 1mg/mL Cd in 0.5N HNO<sub>3</sub>, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cadmium standard solution, for AAS, 1mg/mL Cd in 0.5N HNO3, Thermo Scientific Chemicals
CAS: 10022-68-1 | CdH2N2O6 | 238.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number FSA195875000
Price (MYR)
765.00
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
765.00
Add to cart
Chemical Identifiers
CAS210169-05-4
IUPAC Name5-fluoropyridin-3-amine
Molecular FormulaC5H5FN2
InChI KeyZRORIJXOWXYPMO-UHFFFAOYSA-N
SMILESNC1=CC(F)=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)83.0-90.0?C
Appearance (Color)Pale cream to cream to brown
for atomic adsorption spectroscopy
RUO – Research Use Only