Cadmium standard solution, for AAS, 1mg/mL Cd in 0.5N HNO3, Thermo Scientific Chemicals
Cadmium standard solution, for AAS, 1mg/mL Cd in 0.5N HNO<sub>3</sub>, Thermo Scientific Chemicals
Cadmium standard solution, for AAS, 1mg/mL Cd in 0.5N HNO<sub>3</sub>, Thermo Scientific Chemicals
Cadmium standard solution, for AAS, 1mg/mL Cd in 0.5N HNO<sub>3</sub>, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cadmium standard solution, for AAS, 1mg/mL Cd in 0.5N HNO3, Thermo Scientific Chemicals
CAS: 10022-68-1 | CdH2N2O6 | 238.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number FSA195871000
Price (MYR)
264.00
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
264.00
Add to cart
Chemical Identifiers
CAS10022-68-1, 7697-37-2, 7732-18-5
Specifications
Chemical Name or MaterialCadmium standard solution
Name Note1 mg/ml Cd in 0.5N HNO3
PackagingPlastic bottle
CAS10022-68-1
Quantity100 mL
View more
for atomic adsorption spectroscopy
RUO – Research Use Only