Aluminum standard solution, for AAS, 1 mg/ml Al in 2 to 5% HNO3
Aluminum standard solution, for AAS, 1 mg/ml Al in 2 to 5% HNO3
Aluminum standard solution, for AAS, 1 mg/ml Al in 2 to 5% HNO3
Aluminum standard solution, for AAS, 1 mg/ml Al in 2 to 5% HNO3
Thermo Scientific Chemicals
Aluminum standard solution, for AAS, 1 mg/ml Al in 2 to 5% HNO3
CAS: 7784-27-2 | AlH18N3O18 | 375.129 g/mol
Have Questions?
Catalog NumberQuantity
FSA195805000500 mL
Catalog number FSA195805000
Price (MYR)
2,010.00
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
2,010.00
Add to cart
Chemical Identifiers
CAS3637-01-2
IUPAC Name1-(3,4-dimethylphenyl)ethan-1-one
Molecular FormulaC10H12O
InChI KeyWPRAXAOJIODQJR-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=C(C)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Identification (FTIR)Conforms
Refractive Index1.5370-1.5410 @ 20?C
Assay (GC)≥97.5%
FormLiquid
For atomic adsorption spectroscopy
RUO – Research Use Only