Platinum on activated carbon, 10% Pt, ca .50% moisture, Thermo Scientific Chemicals
Platinum on activated carbon, 10% Pt, ca .50% moisture, Thermo Scientific Chemicals
Platinum on activated carbon, 10% Pt, ca .50% moisture, Thermo Scientific Chemicals
Platinum on activated carbon, 10% Pt, ca .50% moisture, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Platinum on activated carbon, 10% Pt, ca .50% moisture, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
Packaging:
Glass bottle
Catalog number FSA195240100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS112-37-8
IUPAC Nameundecanoic acid
Molecular FormulaC11H22O2
InChI KeyZDPHROOEEOARMN-UHFFFAOYSA-N
SMILESCCCCCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Low melting solid
GC>=98.5 %
Infrared spectrumConforms
Appearance (Color)White to light yellow
Titration with NaOH>=98.5 %