Platinum black, 97+% Pt, Thermo Scientific Chemicals
Platinum black, 97+% Pt, Thermo Scientific Chemicals
Platinum black, 97+% Pt, Thermo Scientific Chemicals
Platinum black, 97+% Pt, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Platinum black, 97+% Pt, Thermo Scientific Chemicals
CAS: 7440-06-4 | Pt | 195.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA195210050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS20666-12-0
IUPAC Namesodium 8-amino-4-oxo-3,4-dihydrophthalazin-1-olate
Molecular FormulaC8H6N3NaO2
InChI KeyGBOXDRMCEVPUSV-UHFFFAOYSA-M
SMILES[Na+].NC1=C2C([O-])=NNC(=O)C2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)> 98.0%
Water Content (Karl Fischer Titration)≤ 1.0%
Sulfated Ash>34.95 to < 36.37% (theory 35.66%)
Appearance (Color)White to cream or pale grey
Assay (ex Sulphated Ash)> 98.0%