Palladium on activated carbon, unreduced, 5% Pd, Thermo Scientific Chemicals
Palladium on activated carbon, unreduced, 5% Pd, Thermo Scientific Chemicals
Palladium on activated carbon, unreduced, 5% Pd, Thermo Scientific Chemicals
Palladium on activated carbon, unreduced, 5% Pd, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Palladium on activated carbon, unreduced, 5% Pd, Thermo Scientific Chemicals
CAS: 7440-05-3 | Pd | 106.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number FSA195020100
Price (MYR)
1,580.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,580.00
Add to cart
Chemical Identifiers
CAS91170-93-3
IUPAC Name3-chloro-4-fluorobenzene-1-sulfonyl chloride
Molecular FormulaC6H3Cl2FO2S
InChI KeyDXFXNSNBZNELII-UHFFFAOYSA-N
SMILESFC1=CC=C(C=C1Cl)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5500-1.5540 @ 20?C
Appearance (Color)Clear yellow
Assay (GC)≥97.5%