Palladium, 99.99%, (trace metal basis), powder, particle size -20 mesh, Thermo Scientific Chemicals
Palladium, 99.99%, (trace metal basis), powder, particle size -20 mesh, Thermo Scientific Chemicals
Palladium, 99.99%, (trace metal basis), powder, particle size -20 mesh, Thermo Scientific Chemicals
Palladium, 99.99%, (trace metal basis), powder, particle size -20 mesh, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Palladium, 99.99%, (trace metal basis), powder, particle size -20 mesh, Thermo Scientific Chemicals
CAS: 7440-05-3 | Pd | 106.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
Packaging:
Glass bottle
Catalog number FSA193690250
Price (MYR)
48,229.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
48,229.00
Add to cart
Chemical Identifiers
CAS7440-25-7
IUPAC Nametantalum
Molecular FormulaTa
InChI KeyGUVRBAGPIYLISA-UHFFFAOYSA-N
SMILES[Ta]
View more
SpecificationsSpecification SheetSpecification Sheet
FormFoil, 1.0mm (0.04in) thick.
Assay from Supplier's CofA99.95% (metals basis)
Appearance (Color)Silvery
Assay from Supplier's CofA≥99.95% (metals basis)
FormFoil, 1.0mm (0.04in) thick, hardened.
For ultra trace analysis
RUO – Research Use Only