Gallium, 99.9999%, (trace metal basis), Thermo Scientific Chemicals
Gallium, 99.9999%, (trace metal basis), Thermo Scientific Chemicals
Gallium, 99.9999%, (trace metal basis), Thermo Scientific Chemicals
Gallium, 99.9999%, (trace metal basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Gallium, 99.9999%, (trace metal basis), Thermo Scientific Chemicals
CAS: 7440-55-3 | Ga | 69.72 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Plastic bottle
Catalog number FSA193070010
Price (MYR)
322.00
Add to cart
Quantity:
1 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
322.00
Add to cart
Chemical Identifiers
CAS1517-69-7
IUPAC Name1-phenylethan-1-ol
Molecular FormulaC8H10O
InChI KeyWAPNOHKVXSQRPX-UHFFFAOYNA-N
SMILESCC(O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5260-1.5300 @ 20°C
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥98.5%
Optical Rotation+45° ± 1° (c=5, methanol)
FormLiquid
View more
For ultra trace analysis
RUO – Research Use Only