Europium(III) oxide, 99.99%, (trace metal basis), Thermo Scientific Chemicals
Europium(III) oxide, 99.99%, (trace metal basis), Thermo Scientific Chemicals
Europium(III) oxide, 99.99%, (trace metal basis), Thermo Scientific Chemicals
Europium(III) oxide, 99.99%, (trace metal basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Europium(III) oxide, 99.99%, (trace metal basis), Thermo Scientific Chemicals
CAS: 1308-96-9 | Eu2O3 | 351.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA193020050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1119-51-3
IUPAC Name5-bromopent-1-ene
Molecular FormulaC5H9Br
InChI KeyLPNANKDXVBMDKE-UHFFFAOYSA-N
SMILESBrCCCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥95.0% (UK-sourced material)
CommentMaterial sourced in UK and US
Assay from Supplier's CofA≥95.0% (US-sourced material)
View more
for ultra trace analysis
RUO – Research Use Only