Bismuth(III) oxide, 98+%, pure
Bismuth(III) oxide, 98+%, pure
Bismuth(III) oxide, 98+%, pure
Bismuth(III) oxide, 98+%, pure
Bismuth(III) oxide, 98+%, pure
CAS: 1304-76-3 | Bi2O3 | 465.96 g/mol
Have Questions?
Catalog number FSA192620050
Price (MYR)
4,841.00
Add to cart
Request bulk or custom format
Price (MYR)
4,841.00
Add to cart
Specifications
Boiling Point1890°C
Chemical Name or MaterialBismuth(III) oxide
Melting Point825.0°C
Name Note99.9999%
CAS1304-76-3
View more
For ultra trace analysis
RUO – Research Use Only