Bismuth(III) oxide, 98+%, pure
Bismuth(III) oxide, 98+%, pure
Bismuth(III) oxide, 98+%, pure
Bismuth(III) oxide, 98+%, pure
Thermo Scientific Chemicals
Bismuth(III) oxide, 98+%, pure
CAS: 1304-76-3 | Bi2O3 | 465.96 g/mol
Have Questions?
Catalog NumberQuantity
FSA1926200101 kg
Catalog number FSA192620010
Price (MYR)
1,223.00
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
1,223.00
Add to cart
Chemical Identifiers
CAS1668-00-4
IUPAC Name3,6-bis[2-(2-arsonophenyl)hydrazin-1-ylidene]-4,5-dioxo-3,4,5,6-tetrahydronaphthalene-2,7-disulfonic acid
Molecular FormulaC22H18As2N4O14S2
InChI KeyTVMZRHVOFZTNET-UHFFFAOYSA-N
SMILESO[As](O)(=O)C1=CC=CC=C1NN=C1C(=O)C2=C(C=C1S(O)(=O)=O)C=C(C(=NNC1=CC=CC=C1[As](O)(O)=O)C2=O)S(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark green to dark brown to dark red to black
Assay from Suppliers CofA≥70%
lambda-max (UV-Vis)530-545nm
FormCrystals or powder or crystalline powder
Loss on Drying≤12.0% (200°C/constant weight)