New Methylene Blue N, zinc chloride double salt, pure
New Methylene Blue N, zinc chloride double salt, pure
New Methylene Blue N, zinc chloride double salt, pure
New Methylene Blue N, zinc chloride double salt, pure
Thermo Scientific Chemicals
New Methylene Blue N, zinc chloride double salt, pure
0·5 ZnCl2
Have Questions?
Catalog NumberQuantity
FSA19202010010 g
Catalog number FSA192020100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS1148-79-4
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)85.5-94.5?C
FormPowder
Assay (GC)≥96.0%
Appearance (Color)Pale cream or pale yellow to orange to brown