Tartrazine, 89%, pure
Tartrazine, 89%, pure
Tartrazine, 89%, pure
Tartrazine, 89%, pure
Thermo Scientific Chemicals
Tartrazine, 89%, pure
CAS: 1934-21-0 | C16H9N4Na3O9S2 | 534.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA191895000
Price (MYR)
765.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
765.00
Add to cart
Chemical Identifiers
CAS107-43-7
IUPAC Name2-(trimethylazaniumyl)acetate
Molecular FormulaC5H11NO2
InChI KeyKWIUHFFTVRNATP-UHFFFAOYSA-N
SMILESC[N+](C)(C)CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Iron (Fe)=<20 ppm
Appearance (Color)White
Titration with HClO497.5 to 102.5 %
Heavy metals (as Pb)=<10 ppm