Tartrazine, 89%, pure
Tartrazine, 89%, pure
Tartrazine, 89%, pure
Tartrazine, 89%, pure
Thermo Scientific Chemicals
Tartrazine, 89%, pure
CAS: 1934-21-0 | C16H9N4Na3O9S2 | 534.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number FSA191891000
Price (MYR)
384.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
384.00
Add to cart
Chemical Identifiers
CAS79604-66-3
IUPAC Name2,4,6-trimethylhepta-1,6-dien-4-ol
Molecular FormulaC10H18O
InChI KeyMETKJWMTLIYYIQ-UHFFFAOYSA-N
SMILESCC(=C)CC(C)(O)CC(C)=C
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4585-1.4625 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥97.5%