Fluorescein diacetate, 97%, pure
Fluorescein diacetate, 97%, pure
Fluorescein diacetate, 97%, pure
Fluorescein diacetate, 97%, pure
Thermo Scientific Chemicals
Fluorescein diacetate, 97%, pure
CAS: 596-09-8 | C24H16O7 | 416.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA191660050
Price (MYR)
403.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
403.00
Add to cart
Chemical Identifiers
CAS335-76-2
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥96.0% (GC and/or titration)
Appearance (Color)White to cream
FormPowder or crystalline powder or Flakes