2-Bromo-6-methoxynaphthalene, 98%, Thermo Scientific Chemicals
2-Bromo-6-methoxynaphthalene, 98%, Thermo Scientific Chemicals
2-Bromo-6-methoxynaphthalene, 98%, Thermo Scientific Chemicals
2-Bromo-6-methoxynaphthalene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Bromo-6-methoxynaphthalene, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA190560050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS87100-15-0
IUPAC Name2-cyclohexyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular FormulaC12H23BO2
InChI KeyOUEVCDGYTKLNMJ-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>96.0%