Sudan II, pure
Sudan II, pure
Sudan II, pure
Thermo Scientific Chemicals
Sudan II, pure
CAS: 3118-97-6 | C18H16N2O | 276.34 g/mol
Have Questions?
Catalog NumberQuantity
FSA19015025025 g
Catalog number FSA190150250
Price (MYR)
493.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
493.00
Add to cart
Chemical Identifiers
CAS107-10-8
IUPAC Namepropan-1-amine
Molecular FormulaC3H9N
InChI KeyWGYKZJWCGVVSQN-UHFFFAOYSA-N
SMILESCCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.2 % (K.F.)
Refractive index1.3870 to 1.3890 (20°C, 589 nm)
Color scale=<20 APHA
Infrared spectrumConforms
GC>=99.0 %