1-Bromooctadecane, 96%, Thermo Scientific Chemicals
1-Bromooctadecane, 96%, Thermo Scientific Chemicals
1-Bromooctadecane, 96%, Thermo Scientific Chemicals
1-Bromooctadecane, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Bromooctadecane, 96%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA190025000
Price (MYR)
1,035.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,035.00
Add to cart
Chemical Identifiers
CAS78-51-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
GC>=94 %
Infrared spectrumConforms
Refractive index1.4360 to 1.4390 (20°C, 589 nm)